Anti-ARRS Mouse Monoclonal Antibody [clone: S128]

Supplier: Biosensis
BSENM-1796-100EA 286 GBP
BSENM-1796-100
Anti-ARRS Mouse Monoclonal Antibody [clone: S128]
Antibodies
Binds to photoactivated, phosphorylated RHO and terminates RHO signaling via G-proteins by competing with G-proteins for the same binding site on RHO (PubMed: 8003967, PubMed: 25205354). May play a role in preventing light-dependent degeneration of retinal photoreceptor cells (By similarity). Ref: uniprot.org. Antibody detects arrestin-1 protein only and does not cross-react with the other 3 arrestin molecules.

Western blotting (1:5,000) and Immunohistochemistry (1:1,000). Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: ARRS
Clonality: Monoclonal
Clone: S128
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR