Anti-PRVA Chicken Polyclonal Antibody

Supplier: Biosensis
BSENC-1814-50EA 286 GBP
BSENC-1814-50
Anti-PRVA Chicken Polyclonal Antibody
Antibodies
In muscle, parvalbumin is thought to be involved in relaxation after contraction. It binds two calcium ions. Ref: uniprot.org. Antibody is specific for parvalbumin and does not recognize closely related proteins calretinin and calbindin as determined by Western Blotting.

Western blotting (1:1,000-1:5,000) and Immunohistochemistry (1:1,000-1:5,000). Note that this antibody does not recognize parvalbumin in rat or mouse brain homogenates on western blots. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: PRVA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR