Northern Ireland
Order Entry
(±)-2-Aminoheptane 98+%
 
 
 
(±)-2-Aminoheptane 98+%
 
undefined
 
 
 
(±)-2-Aminoheptane 98+%
 

 

  • Appearance:
    Clear colorless to yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4175 to 1.4195 (20°C, 589 nm)
  • GC:
    ≥98.0 %

 

 

 

  •  
     
    Pack type
    Pk
     
     
  • ACRO103770250
    103770250
    Glass bottle
    25 mL
       
       
    1 * 25 mL
    1 * 25 mL - £86.50
    £86.50
    1 * 25 mL
       £86.50

     : 103770250
     : ACRO103770250
  • ACRO103771000
    103771000
    Glass bottle
    100 mL
       
       
    1 * 100 mL
    1 * 100 mL - £244.00
    £244.00
    1 * 100 mL
       £244.00

     : 103771000
     : ACRO103771000
  • ACRO103775000
    103775000
    Glass bottle
    500 mL
       
       
    1 * 500 mL
    1 * 500 mL - £927.00
    £927.00
    1 * 500 mL
       £927.00

     : 103775000
     : ACRO103775000