Anti-SIAH1 Mouse Monoclonal Antibody

Supplier: Biosensis
BSENM-867-50EA 315 GBP
BSENM-867-50
Anti-SIAH1 Mouse Monoclonal Antibody
Antibodies
Siah-1 is a member of the seven in absentia homolog (SIAH) family. It is a E3 ubiquitin-protein ligase that mediates ubiquitination and subsequent proteasomal degradation of target proteins, including many proteins involved in transcription regulation, a cell surface receptor (DCC), cytoplasmic signal transduction molecules, an antiapoptotic protein (BAG1), a microtubule motor protein (KIF22), a protein involved in synaptic vesicle function in neurons (SYP) and a structural protein. Consequently Siah-1 is involved in many cellular processes such as apoptosis, tumor suppression, cell cycle, axon guidance, transcription regulation, spermatogenesis, and TNF-alpha signaling. It is predominantly a cytoplasmic protein but is also partially a nuclear protein. It is widely expressed at low levels and is down-regulated in advanced hepatocellular carcinomas.

This antibody is recommended for ELISA. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: SIAH1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR