Copper, Supelco®
M-Clarity™ quality level = MQ100
ECNRM522B-1EAEA
386
GBP
ECNRM522B-1EA
ECNRM522A-1EA
Copper, Supelco®
Copper
Formula:
Cu MW: 63,55 g/mol Boiling Pt: 2595 °C (1013 hPa) Melting Pt: 1083 °C Density: 8,94 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010965 CAS Number: 7440-50-8 EINECS: 231-159-6 Merck Index: 13,02545 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...