Northern Ireland
Order Entry
Anti-UNC45 Mouse Monoclonal Antibody
Anti-UNC45 Mouse Monoclonal Antibody
 
Anti-UNC45 Mouse Monoclonal Antibody

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Catalog No:
  • ENZOADISRA1800D
  • ENZOADISRA1800F
  • Antigen symbol:
  • UNC45
  • UNC45
  • Antigen name:
  • Unc-45 Myosin Chaperone
  • Unc-45 Myosin Chaperone
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1
  • IgG1
  • Western blot:
  • Yes
  • Yes
  • Form:
  • Liquid. In PBS containing 50% glycerol and 0,09% sodium azide.
  • Liquid. In PBS containing 50% glycerol and 0,09% sodium azide.
  • Purification:
  • Protein G affinity purified

 

 

  •  
     
    Pk
     
     
  • ENZOADISRA1800D
    ADISRA1800D
    50 µG
       
       
    1 * 50 µG
    1 * 50 µG - £243.00
    £243.00
    1 * 50 µG
       £243.00

     
     
    Specifications
    Antigen symbol
    UNC45
    Antigen name
    Unc-45 Myosin Chaperone
    Environmentally Preferable
    Antibody type
    Primary
    Clonality
    Monoclonal
    Reactivity
    Human
    Host
    Mouse
    Isotype
    IgG1
    Western blot
    Yes
    Form
    Liquid. In PBS containing 50% glycerol and 0,09% sodium azide.
    Purification
    Protein G affinity purified
  • ENZOADISRA1800F
    ADISRA1800F
    200 µG
       
       
    1 * 200 µG
    1 * 200 µG - £541.00
    £541.00
    1 * 200 µG
       £541.00

     
     
    Specifications
    Antigen symbol
    UNC45
    Antigen name
    Unc-45 Myosin Chaperone
    Environmentally Preferable
    Antibody type
    Primary
    Clonality
    Monoclonal
    Reactivity
    Human
    Host
    Mouse
    Isotype
    IgG1
    Western blot
    Yes
    Form
    Liquid. In PBS containing 50% glycerol and 0,09% sodium azide.
    Purification