Northern Ireland
Order Entry
Anti-HDJ1 Rabbit Polyclonal Antibody
 
Anti-HDJ1 Rabbit Polyclonal Antibody

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Catalog No:
  • ENZOADISPA400F
  • ENZOADISPA400D
  • Antigen symbol:
  • HDJ1
  • HDJ1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Reactivity:
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Form:
  • Liquid. In PBS containing 50% glycerol and 0,09% sodium azide.
  • Liquid. In PBS containing 50% glycerol and 0,09% sodium azide.
  • Purification:
  • Protein A affinity purified
  • Protein A affinity purified

 

 

  •  
     
    Pk
     
     
  • ENZOADISPA400D
    ADISPA400D
    50 µG
       
       
    1 * 50 µG
    1 * 50 µG - £190.00
    £190.00
    1 * 50 µG
       £190.00

     : ADISPA400D
     : ENZOADISPA400D
     
     
    Specifications
    Antigen symbol
    HDJ1
    Environmentally Preferable
    Antibody type
    Primary
    Clonality
    Polyclonal
    Reactivity
    Human
    Host
    Rabbit
    Isotype
    Western blot
    Yes
    ImmunoChemistry
    Yes
    ImmunoPrecipitation
    Yes
    ImmunoFluorescence
    Yes
    Form
    Liquid. In PBS containing 50% glycerol and 0,09% sodium azide.
    Purification
    Protein A affinity purified
  • ENZOADISPA400F
    ADISPA400F
    200 µG
       
       
    1 * 200 µG
    1 * 200 µG - £455.00
    £455.00
    1 * 200 µG
       £455.00

     : ADISPA400F
     : ENZOADISPA400F
     
     
    Specifications
    Antigen symbol
    HDJ1
    Environmentally Preferable
    Antibody type
    Primary
    Clonality
    Polyclonal
    Reactivity
    Host
    Rabbit
    Isotype
    Western blot
    Yes
    ImmunoChemistry
    Yes
    ImmunoPrecipitation
    Yes
    ImmunoFluorescence
    Yes
    Form
    Liquid. In PBS containing 50% glycerol and 0,09% sodium azide.
    Purification
    Protein A affinity purified