Northern Ireland
Order Entry
Anti-GSK3B Rabbit Polyclonal Antibody
Anti-GSK3B Rabbit Polyclonal Antibody
 
Anti-GSK3B Rabbit Polyclonal Antibody

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Catalog No:
  • ENZOADIKAPST002E
  • ENZOADIKAPST002C
  • Antigen symbol:
  • GSK3B
  • GSK3B
  • Antigen name:
  • Glycogen Synthase Kinase 3 beta
  • Glycogen Synthase Kinase 3 beta
  • Antigen synonyms:
  • EC 2.7.11.26|GSK3B|Factor A|GSK-3 beta Factor A|GSK-3 beta
  • EC 2.7.11.26|GSK3B|Factor A|GSK-3 beta Factor A|GSK-3 beta
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Reactivity:
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Form:
  • Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
  • Purification:
  • Protein A affinity purified

 

 

  •  
     
    Size
    Pk
     
     
  • ENZOADIKAPST002C
    ADIKAPST002C
    25 µg
    25 µG
       
       
    1 * 25 µG
    1 * 25 µG - £181.00
    £181.00
    1 * 25 µG
       £181.00

     
     
    Specifications
    Antigen symbol
    GSK3B
    Antigen name
    Glycogen Synthase Kinase 3 beta
    Environmentally Preferable
    Antigen synonyms
    EC 2.7.11.26|GSK3B|Factor A|GSK-3 beta Factor A|GSK-3 beta
    Antibody type
    Primary
    Clonality
    Polyclonal
    Reactivity
    Human
    Host
    Rabbit
    Isotype
    Western blot
    Yes
    ImmunoChemistry
    Yes
    Form
    Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
    Purification
    Protein A affinity purified
  • ENZOADIKAPST002E
    ADIKAPST002E
    100 µg
    100 µG
       
       
    1 * 100 µG
    1 * 100 µG - £390.00
    £390.00
    1 * 100 µG
       £390.00

     
     
    Specifications
    Antigen symbol
    GSK3B
    Antigen name
    Glycogen Synthase Kinase 3 beta
    Environmentally Preferable
    Antigen synonyms
    EC 2.7.11.26|GSK3B|Factor A|GSK-3 beta Factor A|GSK-3 beta
    Antibody type
    Primary
    Clonality
    Polyclonal
    Reactivity
    Host
    Rabbit
    Isotype
    Western blot
    Yes
    ImmunoChemistry
    Yes
    Form
    Purification