Anti-A1 beta Casein Chicken Polyclonal Antibody

Supplier: Biosensis
BSENC-1781-100EA 0 GBP
BSENC-1781-100
Anti-A1 beta Casein Chicken Polyclonal Antibody
Bovine milk contains two types of beta-casein protein, A2 or A1. Recent studies have shown that milk containing the A1 beta casein protein can contribute to issues including gastrointestinal discomfort after ingestion. There is some evidence of a link between ingestion of A1 beta casein protein and the development of Type 1 diabetes.

Type: Primary
Antigen: A1 beta Casein
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope: 78-86
Host: Chicken
Isotype:
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR