Anti-TLR8 Polyclonal Antibody

Supplier: Boster
BSBTRP1073EA 535 GBP
BSBTRP1073
Anti-TLR8 Polyclonal Antibody
Antibodies
Polyclonal antibody for TLR8 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. TLR8 information: Molecular Weight: 119828 MW; Subcellular Localization: Membrane ; Single-pass type I membrane protein ; Tissue Specificity: Detected in brain, heart, lung, liver, placenta, in monocytes, and at lower levels in CD11c+ immature dendritic cells.

Type: Primary
Antigen: TLR8
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR