Engraving device with points
Supplier: Bochem
BOCH12601EA 188 GBP
BOCH12601
Engraving device with points
This engraving device is used for permanent labelling of lab equipment.
  • Engraves metal, glass and plastic
  • Engraving through vibration principle
Delivery information: Engraving device in practical carrying case includes four points.