Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Alloys and Metal Ore Standards

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"9"
  List View Easy View
Sort by:
 
 
 
 

Supplier:  LGC Standards PROMOCHEM
Description:   This standard reference material is in the powder form intended for use in checking chemical methods of analysis and in calibration with instrumental ...
UOM:  1 * 100 g

Supplier:  LGC Standards PROMOCHEM
Description:   This reference standards are intended to use in checking the accuracy of assay methods. These are supplied in fine powder form.
UOM:  1 * 45 g

Supplier:  LGC Standards PROMOCHEM
Description:   GRAY CAST IRON (CARBON + SULFUR) 1 * 150 g
UOM:  1 * 150 g
Catalog Number: (PROONCSDC29114)

Supplier:  LGC Standards PROMOCHEM
Description:   PADRAO MINERIO CHUMBO-ZINCO 1 * 50 g
UOM:  1 * 50 g
Catalog Number: (PROONIST-1764A)

Supplier:  LGC Standards PROMOCHEM
Description:   This reference material is intended for use in the evaluation of chemical and instrumental analysis and in calibration of those methods.
UOM:  1 * 1 items

Supplier:  LGC Standards PROMOCHEM
Description:   CAST IRON (NI-CR-MO) 1 * 150 g
UOM:  1 * 150 g

Supplier:  LGC Standards PROMOCHEM
Description:   LA STEEL (A242) (MOD.) 1 * 1 items
UOM:  1 * 1 items
Catalog Number: (PROONCSHC15601)

Supplier:  LGC Standards PROMOCHEM
Description:   FERROTITANIUM 1 * 50 g
UOM:  1 * 50 g
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is dicontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.
1 - 8  of 9
  1  2  Next 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping