Calibrated electronic thermometers with stainless steel stem
620-0910EA 37.1 GBP
VWRI620-0910
Calibrated electronic thermometers with stainless steel stem
These digital stem thermometers are suitable for field studies, wet lab applications, wash-down areas, food testing and tanks.
  • Reads temperatures in liquids, semi-solids and gases
  • 120 to 197 mm probes easily reach into cuvettes, test tubes, flasks, beakers and more
  • Blunt tip probe is ideal for veterinary practices, farms and ranches
  • Surface probe is ideal for cooking, laboratories and hot plates (does not include probe cover)
  • Features include minimum/maximum temperature readings, alarms, data hold, water resistant models and more
  • Calibration is traceable to NIST
Calibration point: 0; 100 °C
These electronic-stem thermometers are not suitable for solids such as frozen items.
Certifications: Individually serialised, CE marked, ISO/IEC 17025:2005.
Delivery information: Includes accredited calibration documentation and multi-language instructions (English, French, German, Italian, Portuguese and Spanish) plus a LR44 battery.