Anti-SYTL5 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss
BOSSBS-4145R-HRPEA 804 GBP
BOSSBS-4145R-HRP
Anti-SYTL5 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
May act as Rab effector protein and play a role in vesicle trafficking. Binds phospholipids.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: SYTL5
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR