Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody type:Primary
- Antigen name:c-Jun N-terminal kinase 1, c-Jun N-terminal kinase 2, c-Jun N-terminal kinase 3
- Antigen symbol:JNK1 / JNK2 / JNK3
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:26419,26420,26414
- Antigen synonyms:stress-activated protein kinase 1c|MAPK10|JNK3|JNK3A|stress-activated protein kinase 1|JNK1A2|SAPK1a|Stress-activated protein kinase 1b|SAPK1|MAP kinase 10|MAP kinase p49 3F12|Mitogen-activated protein kinase 9|JNK-55|mitogen-activated protein kinase 8 isoform JNK1 alpha1|PRKM9|c-Jun N-terminal kinase 2|PRKM8|JNK-46|SAPK1A|Stress-activated protein kinase JNK3|JNK|MAP kinase 9|SAPK1B|MAPK9|JNK2|MAP kinase 8|JNK1|SAPK1c|SAPK1b|PRKM10|c-Jun N-terminal kinase 1|JUN N-terminal kinase|Stress-activated protein kinase JNK2|MAPK8|MAPK 10|Stress-activated protein kinase 1a|JNK21B1/2|mitogen-activated protein kinase 8 isoform JNK1 beta2|Mitogen-activated protein kinase 10|MAPK 9
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:160-210/384
- Purification:Purified by Protein A
- Pk:100 µl
Serine/threonine-protein kinase involved in various processes such as neuronal proliferation, differentiation, migration and programmed cell death. Extracellular stimuli such as proinflammatory cytokines or physical stress stimulate the stress-activated protein kinase/c-Jun N-terminal kinase (SAP/JNK) signaling pathway. In this cascade, two dual specificity kinases MAP2K4/MKK4 and MAP2K7/MKK7 phosphorylate and activate MAPK1/JNK3. In turn, MAPK1/JNK3 phosphorylates a number of transcription factors, primarily components of AP-1 such as JUN and ATF2 and thus regulates AP-1 transcriptional activity. Plays regulatory roles in the signaling pathways during neuronal apoptosis. Phosphorylates the neuronal microtubule regulator STMN2. Acts in the regulation of the beta-amyloid precursor protein/APP signaling during neuronal differentiation by phosphorylating APP. Participates also in neurite growth in spiral ganglion neurons.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: JNK1 / JNK2 / JNK3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat