Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody type:Primary
- Antigen name:Chemokine (C-C motif) ligand 16
- Antigen symbol:CCL16
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Cross adsorption:No
- Form:liquid
- Gene ID:6360
- Antigen synonyms:NCC-4|SCYL4|ILINCK|CKb12|Mtn-1|LEC|SCYA16|NCC4|HCC-4|LCC-1|LMC
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
LEC (liver expressed cytokine) displays chemotactic activity for lymphocytes and monocytes but not for neutrophils. It also shows a potent myelosuppressive activity and suppresses proliferation of myeloid progenitor cells. Its expression is upregulated by IL10.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CCL16
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human