Anti-LIN28A Mouse Monoclonal Antibody

Supplier: Bioworld Technology
BWRLMB0149EA 368 GBP
BWRLMB0149
Anti-LIN28A Mouse Monoclonal Antibody
Antibodies
Recombinant full length Human LIN28A.

This antibody detects endogenous levels of LIN28A and does not cross-react with related proteins

Type: Primary
Antigen: LIN28A
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR