Full face mask, Panaseal
Supplier: Scott Safety
SCTS1032413EA 234 GBP
SCTS1032413
Full face mask, Panaseal
The Panaseal is a positive pressure facemask that includes neoprene outer mask with reflex seal and neoprene ori-nasal inner mask. The facepiece visor is single curvature and made from a scratch and impact resistant PC material with a flame-retardant coating and provides excellent panoramic vision. The visor is held in place by two U-shaped clamps.
  • Left hand side bayonet connection for the demand valve
  • Option of an additional right quick-fit port to take a communications interface
  • Speech diaphragm at the front of the mask offers unimpeded speech transmission
  • Low resistance positive pressure exhalation valve allows for normal breathing
The mask allows the wearer to wear integrated spectacle frames, without compromising the facemask seal. The mask is designed with separate inspiratory and expiratory paths to minimise the risk of cross contamination of second stage regulators.
Certifications: EN136 Class III; EN137:2006 Class II when used in conjunction with Propak; EN139 when used with Scott airline equipment; AS/NZ 1716