Refractometers with wine scales, OPTi
Supplier: Bellingham + Stanley
635-0605EA 328 GBP
BELL38-47 BELL38-41 BELL38-42 BELL38-43 BELL38-44 BELL38-45 BELL38-46
Refractometers with wine scales, OPTi
The OPTi are quality instruments designed for applications in wine testing.
  • Shallow prism dish made of 316 stainless steel
  • Zero calibration with distilled water
  • Automated temperature compensation (ATC)
Delivery information: Supplied with two batteries, a multi-language guide, a certificate of calibration in a protective case.