Centrifuges of choice in development and quality control laboratories of the life science industry and in medical routine laboratories.
- Smooth and quiet running, exceptional speed stability
- Frequency-controlled drive, maintenance only required according to UVV regulations, nine acceleration/braking ramps with extremely short run-up and run-down times
- 89 programmable memories
- Safety features: Panelling with scratch and impact resistant coating, automatic lid locking and holding, lid dropping protection, emergency lid lock release, automatic rotor recognition, imbalance switch-off, key-operated switch for locking and protecting programs, motor overheating protection, chamber overheating protection for ROTIXA 50 RS
- Options: Centrifuging data report system HettInfo (for easy data documentation); Barcode reader; Visual indication when the rotor is stationary; Chaining of programs (allowing to program the centrifuge to run different programs in succession); Second independent tacho system for calibrating the speed measuring instrument as required by quality management systems
Suited for separating a wide range of samples such as food and beverage, cell suspensions, sediments of chemical reactions, sludge and, of course, blood and blood components.
Ordering information: Rotors and adapters available separately.