Gas analysis system
Supplier: Testo
TEST0632.3511EA 0 GBP
TEST0632.3511 TEST0632.3510
Gas analysis system
This gas analyser is designed to control emission measurements of different measuring and analysis applications, also suited for complex data compilation.
  • Four different applications: Burner, gas turbine, motors and user-defined applications
  • Instructions on the display make the steps leading up to the start of the measurement easier
  • Especially advisable when carrying out work on larger systems
Can also be used to remote control the Analysis Box when it is not positioned at the exhaust gas pipe or measurement site.
Certifications: IP 40
Delivery information: Comes with an oxygen gas sensor and a rechargeable battery. You will need to connect at least one sensor (maximum six) to operate the box. Optional sensors include gas sensors for CO, CO₂, NO, NO₂, SO₂, H₂S and CxHy.
Analyser box, equipped with O₂ differential pressure sensor, temperature probe input Type K NiCr-Ni and Type S Pt10Rh-Pt, connection databus, rechargeable battery, integrated combustion air probe (NTC), trigger input, measurement data store and USB interface. Optional sensors include gas sensors for CO, CO₂, NO, NO₂, SO₂, H₂S and CxHy.