Bottle holder, silicone, DURAN®
Supplier: DWK Life Sciences
SCOT292135401EA 0 GBP
SCOT292135401
Bottle holder, silicone, DURAN®
The silicone bottle holder features eight flexible support ribs and stabilises bottles and bottle systems when dispensing, pipetting or filtering, for example.
  • Excellent temperature resistance
  • Excellent chemical resistance
  • Easy to clean
The bottle holder centres the centre of gravity of a laboratory glass bottle or laboratory glass bottle system. The bottle is held securely in the non-slip holder, allowing staff to work conveniently and safely.