Northern Ireland
Order Entry
Anti-S100A1 Mouse Monoclonal Antibody (Biotin) [clone: S100A1/1012]
Anti-S100A1 Mouse Monoclonal Antibody (Biotin) [clone: S100A1/1012]
 :  Biotium
 
Anti-S100A1 Mouse Monoclonal Antibody (Biotin) [clone: S100A1/1012]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNCB1012-100
  • BNCB1012-500
  • Antigen symbol:
  • S100A1
  • S100A1
  • Antigen name:
  • S100
  • S100
  • Antigen synonyms:
  • S100 Calcium Binding Protein A1|Bpb|S100 Alpha Chain|S100 Calcium Binding Protein B|NEF|S100-A1|S100 Beta Chain|S100 Calcium Binding Protein Beta Neural
  • S100 Calcium Binding Protein A1|Bpb|S100 Alpha Chain|S100 Calcium Binding Protein B|NEF|S100-A1|S100 Beta Chain|S100 Calcium Binding Protein Beta Neural
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Clone:
  • Clone S100A1/1012
  • Clone S100A1/1012
  • Reactivity:
  • Human, Bovine, Rat, Mouse
  • Human, Bovine, Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 515715
  • 515715
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Product Information

  • Catalog #
    Supplier #
    Size
    Pk
    Availability
    Price
  • BNCB1012-100
    BNCB1012-100
    100 μL
    100 µl
       
    Log in for availability
    1 * 100 µl
    1 * 100 µl - £195.00
    £195.00
    1 * 100 µl
       £195.00

     
     
    Specifications
    Antigen symbol
    S100A1
    Antigen name
    S100
    Environmentally Preferable
    Antigen synonyms
    S100 Calcium Binding Protein A1|Bpb|S100 Alpha Chain|S100 Calcium Binding Protein B|NEF|S100-A1|S100 Beta Chain|S100 Calcium Binding Protein Beta Neural
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    Biotin
    Clone
    Clone S100A1/1012
    Reactivity
    Human, Bovine, Rat, Mouse
    Host
    Mouse
    Gene ID
    515715
    Isotype
    IgG2a kappa
    Western blot
    Yes
    ImmunoChemistry
    Yes
    ImmunoFluorescence
    Yes
    Concentration
    0,1 mg/ml
    Storage buffer
    PBS/0,1% BSA/0,05% azide
  • BNCB1012-500
    BNCB1012-500
    500 μL
    500 µl
       
       
    1 * 500 µl
    1 * 500 µl - £538.00
    £538.00
    1 * 500 µl
       £538.00

     
     
    Specifications
    Antigen symbol
    S100A1
    Antigen name
    S100
    Environmentally Preferable
    Antigen synonyms
    S100 Calcium Binding Protein A1|Bpb|S100 Alpha Chain|S100 Calcium Binding Protein B|NEF|S100-A1|S100 Beta Chain|S100 Calcium Binding Protein Beta Neural
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    Biotin
    Clone
    Clone S100A1/1012
    Reactivity
    Human, Bovine, Rat, Mouse
    Host
    Mouse
    Gene ID
    515715
    Isotype
    IgG2a kappa
    Western blot
    Yes
    ImmunoChemistry
    Yes
    ImmunoFluorescence
    Yes
    Concentration
    0,1 mg/ml
    Storage buffer
    PBS/0,1% BSA/0,05% azide