Northern Ireland
Order Entry
Anti-KRT8 & KRT18 Mouse Monoclonal Antibody [clone: KRT8/803 KRT18/835]
 :  Biotium
 
Anti-KRT8 & KRT18 Mouse Monoclonal Antibody [clone: KRT8/803 KRT18/835]
 :  Biotium

 

  • Catalog No:
  • BNUB0979-100
  • BNUB0979-500
  • Antigen symbol:
  • KRT8 & KRT18
  • KRT8 & KRT18
  • Antigen name:
  • Cytokeratin 8/18
  • Cytokeratin 8/18
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • Clone KRT8/803 KRT18/835
  • Clone KRT8/803 KRT18/835
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 533782 , 708445 (CK8); 406013 (CK18)
  • 533782 , 708445 (CK8); 406013 (CK18)
  • Isotype:
  • IgGs
  • IgGs
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,2 mg/ml
  • 0,2 mg/ml
  • Storage buffer:
  • PBS/0,05% BSA/0,05% azide
  • PBS/0,05% BSA/0,05% azide

 

 

  •  
     
    Size
    Pk
     
     
  • BNUB0979-100
    BNUB0979-100
    100 μL
    100 µl
       
       
    1 * 100 µl
    1 * 100 µl - £228.00
    £228.00
    1 * 100 µl
       £228.00

     
     
    Specifications
    Antigen symbol
    KRT8 & KRT18
    Antigen name
    Cytokeratin 8/18
    Environmentally Preferable
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    Unconjugated
    Clone
    Clone KRT8/803 KRT18/835
    Reactivity
    Human
    Host
    Mouse
    Gene ID
    533782 , 708445 (CK8); 406013 (CK18)
    Isotype
    IgGs
    Western blot
    Yes
    ImmunoChemistry
    Yes
    ImmunoFluorescence
    Yes
    Concentration
    0,2 mg/ml
    Storage buffer
    PBS/0,05% BSA/0,05% azide
  • BNUB0979-500
    BNUB0979-500
    500 μL
    500 µl
       
       
    1 * 500 µl
    1 * 500 µl - £491.00
    £491.00
    1 * 500 µl
       £491.00

     
     
    Specifications
    Antigen symbol
    KRT8 & KRT18
    Antigen name
    Cytokeratin 8/18
    Environmentally Preferable
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    Unconjugated
    Clone
    Clone KRT8/803 KRT18/835
    Reactivity
    Human
    Host
    Mouse
    Gene ID
    533782 , 708445 (CK8); 406013 (CK18)
    Isotype
    IgGs
    Western blot
    Yes
    ImmunoChemistry
    Yes
    ImmunoFluorescence
    Yes
    Concentration
    0,2 mg/ml
    Storage buffer
    PBS/0,05% BSA/0,05% azide