Northern Ireland
Order Entry
Anti-CD7 Mouse Monoclonal Antibody (Purified) [clone: T3-3A1]
Supplier:  Biotium
Certificates
Anti-CD7 Mouse Monoclonal Antibody (Purified) [clone: T3-3A1]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNUB0978-100
  • BNUB0978-500
  • Antigen symbol:
  • CD7
  • CD7
  • Antigen synonyms:
  • TP41|Leu9|T-cell surface antigen Leu-9|T-cell leukemia antigen|Tp40|p41|GP40
  • TP41|Leu9|T-cell surface antigen Leu-9|T-cell leukemia antigen|Tp40|p41|GP40
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • Clone T3-3A1
  • Clone T3-3A1
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 186820
  • 186820
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,2 mg/ml
  • 0,2 mg/ml
  • Storage buffer:
  • PBS/0,05% BSA/0,05% azide
  • PBS/0,05% BSA/0,05% azide

Specifications

Product Information

  • Catalog #
    Supplier #
    Size
    Pk
    Availability
    Price
  • BNUB0978-100
    BNUB0978-100
    100 μL
    100 µl
       
    Log in for availability
    1 * 100 µl
    1 * 100 µl - £228.00
    £228.00
    1 * 100 µl
       £228.00

     : BNUB0978-100
     : BTIUBNUB0978-100
    Specifications:
    More
    Specifications
    Antigen symbol
    CD7
    Environmentally Preferable
    Antigen synonyms
    TP41|Leu9|T-cell surface antigen Leu-9|T-cell leukemia antigen|Tp40|p41|GP40
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    Unconjugated
    Clone
    Clone T3-3A1
    Reactivity
    Human
    Host
    Mouse
    Gene ID
    186820
    Isotype
    IgG1 kappa
    ImmunoFluorescence
    Yes
    Concentration
    0,2 mg/ml
    Storage buffer
    PBS/0,05% BSA/0,05% azide
  • BNUB0978-500
    BNUB0978-500
    500 μL
    500 µl
       
       
    1 * 500 µl
    1 * 500 µl - £491.00
    £491.00
    1 * 500 µl
       £491.00

     : BNUB0978-500
     : BTIUBNUB0978-500
     
     
    Specifications
    Antigen symbol
    CD7
    Environmentally Preferable
    Antigen synonyms
    TP41|Leu9|T-cell surface antigen Leu-9|T-cell leukemia antigen|Tp40|p41|GP40
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    Unconjugated
    Clone
    Clone T3-3A1
    Reactivity
    Human
    Host
    Mouse
    Gene ID
    186820
    Isotype
    IgG1 kappa
    ImmunoFluorescence
    Yes
    Concentration
    0,2 mg/ml
    Storage buffer
    PBS/0,05% BSA/0,05% azide