Northern Ireland
Order Entry
Anti-UCHL1 Rabbit Polyclonal Antibody
 
undefined
Anti-UCHL1 Rabbit Polyclonal Antibody

 

  • Catalog No:
  • NOVUNB300-676SS
  • NOVUNB300-676
  • Antigen symbol:
  • UCHL1
  • UCHL1
  • Antigen name:
  • Ubiquitin Carboxyl-Terminal Esterase L1
  • Ubiquitin Carboxyl-Terminal Esterase L1
  • Antigen synonyms:
  • Neuron cytoplasmic protein 9.5|Ubiquitin thioesterase L1.|PGP9.5|Ubiquitin carboxyl-terminal|hydrolase isozyme L1|Ubiquitin thioesterase L1|UCH-L1|PGP 9.5|PARK5
  • Neuron cytoplasmic protein 9.5|Ubiquitin thioesterase L1.|PGP9.5|Ubiquitin carboxyl-terminal|hydrolase isozyme L1|Ubiquitin thioesterase L1|UCH-L1|PGP 9.5|PARK5
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 7345
  • 7345
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

 

 

 

  •  
     
    Size
    Pk
     
     
  • NOVUNB300-676SS
    NB300-676SS
    25 μL
    25 µl
       
       
    1 * 25 µl
    1 * 25 µl - £198.00
    £198.00
    1 * 25 µl
       £198.00

     
     
    Specifications
    Antigen symbol
    UCHL1
    Antigen name
    Ubiquitin Carboxyl-Terminal Esterase L1
    Environmentally Preferable
    Antigen synonyms
    Neuron cytoplasmic protein 9.5|Ubiquitin thioesterase L1.|PGP9.5|Ubiquitin carboxyl-terminal|hydrolase isozyme L1|Ubiquitin thioesterase L1|UCH-L1|PGP 9.5|PARK5
    Antibody type
    Primary
    Clonality
    Polyclonal
    Conjugation
    Unconjugated
    Reactivity
    Human
    Host
    Rabbit
    Gene ID
    7345
    Isotype
    Western blot
    Yes
    ImmunoChemistry
    Yes
    ImmunoFluorescence
    Yes
    Storage temperature
    Cool
    Shipping temperature
    Ice
  • NOVUNB300-676
    NB300-676
    100 μL
    0,1 mL
       
       
    1 * 0,1 mL
    1 * 0,1 mL - £509.00
    £509.00
    1 * 0,1 mL
       £509.00

     
     
    Specifications
    Antigen symbol
    UCHL1
    Antigen name
    Ubiquitin Carboxyl-Terminal Esterase L1
    Environmentally Preferable
    Antigen synonyms
    Neuron cytoplasmic protein 9.5|Ubiquitin thioesterase L1.|PGP9.5|Ubiquitin carboxyl-terminal|hydrolase isozyme L1|Ubiquitin thioesterase L1|UCH-L1|PGP 9.5|PARK5
    Antibody type
    Primary
    Clonality
    Polyclonal
    Conjugation
    Unconjugated
    Reactivity
    Human
    Host
    Rabbit
    Gene ID
    7345
    Isotype
    Western blot
    Yes
    ImmunoChemistry
    Yes
    ImmunoFluorescence
    Yes
    Storage temperature
    Cool
    Shipping temperature
    Ice