Northern Ireland
Order Entry
Anti-MITF Mouse Monoclonal Antibody (CF647) [clone: D5]
 :  Biotium
 
Anti-MITF Mouse Monoclonal Antibody (CF647) [clone: D5]
 :  Biotium

 

  • Catalog No:
  • BNC470892-500
  • BNC470892-100
  • Antigen symbol:
  • MITF
  • MITF
  • Antigen name:
  • MiTF
  • MiTF
  • Antigen synonyms:
  • CMM8|BHLHE32|MITF|Class E basic helix-loop-helix protein 32 (bHLHe32)|WS2|WS2A|Microphthalmia-associated transcription factor|Mi
  • CMM8|BHLHE32|MITF|Class E basic helix-loop-helix protein 32 (bHLHe32)|WS2|WS2A|Microphthalmia-associated transcription factor|Mi
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 647
  • CF® 647
  • Clone:
  • Clone D5
  • Clone D5
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 166017 , 618266
  • 166017 , 618266
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

 

 

  •  
     
    Size
    Pk
     
     
  • BNC470892-500
    BNC470892-500
    500 μL
    500 µl
       
       
    1 * 500 µl
    1 * 500 µl - £538.00
    £538.00
    1 * 500 µl
       £538.00

     
     
    Specifications
    Antigen symbol
    MITF
    Antigen name
    MiTF
    Environmentally Preferable
    Antigen synonyms
    CMM8|BHLHE32|MITF|Class E basic helix-loop-helix protein 32 (bHLHe32)|WS2|WS2A|Microphthalmia-associated transcription factor|Mi
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    CF® 647
    Clone
    Clone D5
    Reactivity
    Human
    Host
    Mouse
    Gene ID
    166017 , 618266
    Isotype
    IgG1 kappa
    ImmunoChemistry
    Yes
    ImmunoFluorescence
    Yes
    Concentration
    0,1 mg/ml
    Storage buffer
    PBS/0,1% BSA/0,05% azide
  • BNC470892-100
    BNC470892-100
    100 μL
    100 µl
       
       
    1 * 100 µl
    1 * 100 µl - £195.00
    £195.00
    1 * 100 µl
       £195.00

     
     
    Specifications
    Antigen symbol
    MITF
    Antigen name
    MiTF
    Environmentally Preferable
    Antigen synonyms
    CMM8|BHLHE32|MITF|Class E basic helix-loop-helix protein 32 (bHLHe32)|WS2|WS2A|Microphthalmia-associated transcription factor|Mi
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    CF® 647
    Clone
    Clone D5
    Reactivity
    Human
    Host
    Mouse
    Gene ID
    166017 , 618266
    Isotype
    IgG1 kappa
    ImmunoChemistry
    Yes
    ImmunoFluorescence
    Yes
    Concentration
    0,1 mg/ml
    Storage buffer
    PBS/0,1% BSA/0,05% azide