This MAb reacts specifically with heat shock protein HSP27 in human and monkey tissues and cell lines such as MCF-7. HSP27, also referred to as the Estrogen-Regulated 24K protein and HSP28, is one of several small heat shock proteins produced by all organisms studied. HSP27 synthesis is induced by elevated temperature, as well as by estrogen in hormone responsive cells. Interestingly, human HSP27 also shares greater than 50% homology with low molecular weight Drosophila HSPs and mammalian α-crystalline lens protein. Because of the estrogen responsive nature of HSP27, this protein has been studied extensively in human estrogen responsive tissues such as cervix, endometrium and breast tissue. Therefore HSP27 may be useful in classifying various hormone sensitive tumors.
Type: Primary
Antigen: HSPB1
Clonality: Monoclonal
Clone: G3.1
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Chicken, Chimpanzee, Human, Monkey, Mouse, Rat, Sheep
- Catalog No:
- BNC400861-500
- BTIUBNC400861-100
- Antigen symbol:
- HSPB1
- HSPB1
- Antigen name:
- HSP27
- HSP27
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 640R
- CF® 640R
- Clone:
- Clone G3.1
- Clone G3.1
- Reactivity:
- Monkey, Sheep, Chicken, Chimpanzee, Human, Rat, Mouse
- Monkey, Sheep, Chicken, Chimpanzee, Human, Rat, Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- 520973
- 520973
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide
- SizePk
Specifications
Antigen symbolHSPB1Antigen nameHSP27Environmentally PreferableAntibody typePrimaryClonalityMonoclonalConjugationCF® 640RCloneClone G3.1ReactivityMonkey, Sheep, Chicken, Chimpanzee, Human, Rat, MouseHostMouseGene ID520973IsotypeIgG1 kappaWestern blotYesImmunoChemistryYesImmunoFluorescenceYesConcentration0,1 mg/mlStorage bufferPBS/0,1% BSA/0,05% azide- BTIUBNC400861-100BNC400861-100100 μl100 µl1 * 100 µl£195.00
Specifications
Antigen symbolHSPB1Antigen nameHSP27Environmentally PreferableAntibody typePrimaryClonalityMonoclonalConjugationCF® 640RCloneClone G3.1ReactivityMonkey, Sheep, Chicken, Chimpanzee, Human, Rat, MouseHostMouseGene ID520973IsotypeIgG1 kappaWestern blotYesImmunoChemistryYesImmunoFluorescenceYesConcentration0,1 mg/mlStorage bufferPBS/0,1% BSA/0,05% azide