Northern Ireland
Order Entry
Anti-HSPB1 Mouse Monoclonal Antibody (CF488A) [clone: G3.1]
Anti-HSPB1 Mouse Monoclonal Antibody (CF488A) [clone: G3.1]
 :  Biotium
 
Anti-HSPB1 Mouse Monoclonal Antibody (CF488A) [clone: G3.1]
 :  Biotium

 

  • Catalog No:
  • BNC880861-500
  • BNC880861-100
  • Antigen symbol:
  • HSPB1
  • HSPB1
  • Antigen name:
  • HSP27
  • HSP27
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 488A
  • CF® 488A
  • Clone:
  • Clone G3.1
  • Clone G3.1
  • Reactivity:
  • Monkey, Sheep, Chicken, Chimpanzee, Human, Rat, Mouse
  • Monkey, Sheep, Chicken, Chimpanzee, Human, Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 520973
  • 520973
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

 

 

  •  
     
    Size
    Pk
     
     
  • BNC880861-500
    BNC880861-500
    500 μl
    500 µl
       
       
    1 * 500 µl
    1 * 500 µl - £538.00
    £538.00
    1 * 500 µl
       £538.00

     
     
    Specifications
    Antigen symbol
    HSPB1
    Antigen name
    HSP27
    Environmentally Preferable
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    CF® 488A
    Clone
    Clone G3.1
    Reactivity
    Monkey, Sheep, Chicken, Chimpanzee, Human, Rat, Mouse
    Host
    Mouse
    Gene ID
    520973
    Isotype
    IgG1 kappa
    Western blot
    Yes
    ImmunoChemistry
    Yes
    ImmunoFluorescence
    Yes
    Concentration
    0,1 mg/ml
    Storage buffer
    PBS/0,1% BSA/0,05% azide
  • BNC880861-100
    BNC880861-100
    100 μl
    100 µl
       
       
    1 * 100 µl
    1 * 100 µl - £195.00
    £195.00
    1 * 100 µl
       £195.00

     
     
    Specifications
    Antigen symbol
    HSPB1
    Antigen name
    HSP27
    Environmentally Preferable
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    CF® 488A
    Clone
    Clone G3.1
    Reactivity
    Monkey, Sheep, Chicken, Chimpanzee, Human, Rat, Mouse
    Host
    Mouse
    Gene ID
    520973
    Isotype
    IgG1 kappa
    Western blot
    Yes
    ImmunoChemistry
    Yes
    ImmunoFluorescence
    Yes
    Concentration
    0,1 mg/ml
    Storage buffer
    PBS/0,1% BSA/0,05% azide