Northern Ireland
Order Entry
Anti-BCL2 Mouse Monoclonal Antibody (Biotin) [clone: BCL2/782]
Anti-BCL2 Mouse Monoclonal Antibody (Biotin) [clone: BCL2/782]
 :  Biotium
 
Anti-BCL2 Mouse Monoclonal Antibody (Biotin) [clone: BCL2/782]
 :  Biotium

 

  • Catalog No:
  • BNCB0782-100
  • BNCB0782-500
  • Antigen symbol:
  • BCL2
  • BCL2
  • Antigen name:
  • bcl-2
  • bcl-2
  • Antigen synonyms:
  • Apoptosis regulator Bcl-2|B-cell CLL/lymphoma-2
  • Apoptosis regulator Bcl-2|B-cell CLL/lymphoma-2
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Clone:
  • Clone BCL2/782
  • Clone BCL2/782
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 150749
  • 150749
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

 

 

  •  
     
    Size
    Pk
     
     
  • BNCB0782-100
    BNCB0782-100
    100 μL
    100 µl
       
       
    1 * 100 µl
    1 * 100 µl - £195.00
    £195.00
    1 * 100 µl
       £195.00

     : BNCB0782-100
     : BTIUBNCB0782-100
     
     
    Specifications
    Antigen symbol
    BCL2
    Antigen name
    bcl-2
    Environmentally Preferable
    Antigen synonyms
    Apoptosis regulator Bcl-2|B-cell CLL/lymphoma-2
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    Biotin
    Clone
    Clone BCL2/782
    Reactivity
    Human
    Host
    Mouse
    Gene ID
    150749
    Isotype
    IgG1 kappa
    Western blot
    Yes
    ImmunoChemistry
    Yes
    ImmunoFluorescence
    Yes
    Concentration
    0,1 mg/ml
    Storage buffer
    PBS/0,1% BSA/0,05% azide
  • BNCB0782-500
    BNCB0782-500
    500 μL
    500 µl
       
       
    1 * 500 µl
    1 * 500 µl - £538.00
    £538.00
    1 * 500 µl
       £538.00

     : BNCB0782-500
     : BTIUBNCB0782-500
     
     
    Specifications
    Antigen symbol
    BCL2
    Antigen name
    bcl-2
    Environmentally Preferable
    Antigen synonyms
    Apoptosis regulator Bcl-2|B-cell CLL/lymphoma-2
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    Biotin
    Clone
    Clone BCL2/782
    Reactivity
    Human
    Host
    Mouse
    Gene ID
    150749
    Isotype
    IgG1 kappa
    Western blot
    Yes
    ImmunoChemistry
    Yes
    ImmunoFluorescence
    Yes
    Concentration
    0,1 mg/ml
    Storage buffer
    PBS/0,1% BSA/0,05% azide