This MAb reacts with a 200 kDa and 68 kDa protein, identified as heavy and light sub-units of neurofilaments (NF-H & NF-L). Neurofilaments make up the main structural elements of axons and dendrites and are found in neurons, peripheral nerves, and sympathetic ganglion cells. Neurofilaments consist of three major subunits with molecular weights of 68 kDa (NF-L), 160 kDa (NF-M) and 200 kDa (NF-H). Anti-neurofilament stains a number of neural, neuroendocrine, and endocrine tumors. Neuromas, ganglioneuromas, gangliogliomas, ganglioneuroblastomas, and neuroblastomas stain positively for anti-neurofilament. Neurofilaments are also present in paragangliomas as well as adrenal and extra-adrenal pheochromocytomas. Carcinoids, neuroendocrine carcinomas of the skin, and oat cell carcinomas of the lung also express neurofilament.
Type: Primary
Antigen: NEFH & NEFL
Clonality: Monoclonal
Clone: NF421 NFL/736
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Chicken, Human, Mouse, Pig, Rat
- Catalog No:
- BNC940756-100
- BNC940756-500
- Antigen symbol:
- NEFH & NEFL
- NEFH & NEFL
- Antigen name:
- Neurofilament
- Neurofilament
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 594
- CF® 594
- Clone:
- Clone NF421 NFL/736
- Clone NF421 NFL/736
- Reactivity:
- Pig, Chicken, Human, Rat, Mouse
- Pig, Chicken, Human, Rat, Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- 198760
- 198760
- Isotype:
- IgG
- IgG
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide
- SizePk
- : BNC940756-100: BTIUBNC940756-100
Specifications
Antigen symbolNEFH & NEFLAntigen nameNeurofilamentEnvironmentally PreferableAntibody typePrimaryClonalityMonoclonalConjugationCF® 594CloneClone NF421 NFL/736ReactivityPig, Chicken, Human, Rat, MouseHostMouseGene ID198760IsotypeIgGImmunoChemistryYesImmunoFluorescenceYesConcentration0,1 mg/mlStorage bufferPBS/0,1% BSA/0,05% azide - : BNC940756-500: BTIUBNC940756-500
Specifications
Antigen symbolNEFH & NEFLAntigen nameNeurofilamentEnvironmentally PreferableAntibody typePrimaryClonalityMonoclonalConjugationCF® 594CloneClone NF421 NFL/736ReactivityPig, Chicken, Human, Rat, MouseHostMouseGene ID198760IsotypeIgGImmunoChemistryYesImmunoFluorescenceYesConcentration0,1 mg/mlStorage bufferPBS/0,1% BSA/0,05% azide