Anti-NPY5R Rabbit Polyclonal Antibody

Supplier: Bioss
BOSSBS-11521REA 326 GBP
BOSSBS-11521R
Anti-NPY5R Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NPY5R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR