Northern Ireland
Order Entry
Anti-KRT8 & KRT18 Mouse Monoclonal Antibody (CF405S) [clone: K8.8 DC10]
Anti-KRT8 & KRT18 Mouse Monoclonal Antibody (CF405S) [clone: K8.8 DC10]
 :  Biotium
 
Anti-KRT8 & KRT18 Mouse Monoclonal Antibody (CF405S) [clone: K8.8 DC10]
 :  Biotium

 

  • Catalog No:
  • BNC040691-100
  • BNC040691-500
  • Antigen symbol:
  • KRT8 & KRT18
  • KRT8 & KRT18
  • Antigen name:
  • Cytokeratin 8/18
  • Cytokeratin 8/18
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 405S
  • CF® 405S
  • Clone:
  • Clone K8.8 DC10
  • Clone K8.8 DC10
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 533782 , 708445 (CK8); 406013 (CK18)
  • 533782 , 708445 (CK8); 406013 (CK18)
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

 

 

  •  
     
    Size
    Pk
     
     
  • BNC040691-100
    BNC040691-100
    100 μL
    100 µl
       
       
    1 * 100 µl
    1 * 100 µl - £195.00
    £195.00
    1 * 100 µl
       £195.00

     
     
    Specifications
    Antigen symbol
    KRT8 & KRT18
    Antigen name
    Cytokeratin 8/18
    Environmentally Preferable
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    CF® 405S
    Clone
    Clone K8.8 DC10
    Reactivity
    Human
    Host
    Mouse
    Gene ID
    533782 , 708445 (CK8); 406013 (CK18)
    Isotype
    IgG1 kappa
    Western blot
    Yes
    ImmunoChemistry
    Yes
    ImmunoFluorescence
    Yes
    Concentration
    0,1 mg/ml
    Storage buffer
    PBS/0,1% BSA/0,05% azide
  • BNC040691-500
    BNC040691-500
    500 μL
    500 µl
       
       
    1 * 500 µl
    1 * 500 µl - £538.00
    £538.00
    1 * 500 µl
       £538.00

     
     
    Specifications
    Antigen symbol
    KRT8 & KRT18
    Antigen name
    Cytokeratin 8/18
    Environmentally Preferable
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    CF® 405S
    Clone
    Clone K8.8 DC10
    Reactivity
    Human
    Host
    Mouse
    Gene ID
    533782 , 708445 (CK8); 406013 (CK18)
    Isotype
    IgG1 kappa
    Western blot
    Yes
    ImmunoChemistry
    Yes
    ImmunoFluorescence
    Concentration
    0,1 mg/ml
    Storage buffer
    PBS/0,1% BSA/0,05% azide