Anti-MC5R Rabbit Polyclonal Antibody

Supplier: Bioss
BOSSBS-11418REA 648 GBP
BOSSBS-11418R
Anti-MC5R Rabbit Polyclonal Antibody
Antibodies
Receptor for MSH (alpha, beta and gamma) and ACTH. The activity of this receptor is mediated by G proteins which activate adenylate cyclase. This receptor is a possible mediator of the immunomodulation properties of melanocortins.

Recommended Dilutions: Western Blot: 1:100 to 1000; Flow Cytometry: 1:20 to 100; IHC-P: 1:100 to 500; IF(IHC-P): 1:50 to 200.

Type: Primary
Antigen: MC5R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR