Northern Ireland
Order Entry
Anti-CD2 Mouse Monoclonal Antibody (Purified) [clone: LFA2/600]
Anti-CD2 Mouse Monoclonal Antibody (Purified) [clone: LFA2/600]
 :  Biotium
 
Anti-CD2 Mouse Monoclonal Antibody (Purified) [clone: LFA2/600]
Supplier:  Biotium

 

  • Catalog No:
  • BNUB0600-100
  • BNUB0600-500
  • Antigen symbol:
  • CD2
  • CD2
  • Antigen name:
  • CD2 molecule
  • CD2 molecule
  • Antigen synonyms:
  • Rosette receptor|Erythrocyte receptor|Ly-37|T-cell surface antigen T11/Leu-5|T11|T-cell surface antigen CD2|SRBC|LFA-3 receptor|LFA-2|Lymphocyte function antigen 2
  • Rosette receptor|Erythrocyte receptor|Ly-37|T-cell surface antigen T11/Leu-5|T11|T-cell surface antigen CD2|SRBC|LFA-3 receptor|LFA-2|Lymphocyte function antigen 2
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • Clone LFA2/600
  • Clone LFA2/600
  • Reactivity:
  • Primates, Pig, Human
  • Primates, Pig, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 523500
  • 523500
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoFluorescence:
  • Yes
  • Concentration:
  • 0,2 mg/ml
  • 0,2 mg/ml
  • Storage buffer:
  • PBS/0,05% BSA/0,05% azide
  • PBS/0,05% BSA/0,05% azide

 

Product Information

  • Catalog #
    Supplier #
    Size
    Pk
    Availability
    Price
  • BNUB0600-100
    BNUB0600-100
    100 μL
    100 µl
       
    Log in for availability
    1 * 100 µl
    1 * 100 µl - £228.00
    £228.00
    1 * 100 µl
       £228.00

    Supplier Article Number: BNUB0600-100
    Local Article Number: BTIUBNUB0600-100
     
     
    Specifications
    Antigen symbol
    CD2
    Antigen name
    CD2 molecule
    Environmentally Preferable
    Antigen synonyms
    Rosette receptor|Erythrocyte receptor|Ly-37|T-cell surface antigen T11/Leu-5|T11|T-cell surface antigen CD2|SRBC|LFA-3 receptor|LFA-2|Lymphocyte function antigen 2
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    Unconjugated
    Clone
    Clone LFA2/600
    Reactivity
    Primates, Pig, Human
    Host
    Mouse
    Gene ID
    523500
    Isotype
    IgG1 kappa
    ImmunoFluorescence
    Concentration
    0,2 mg/ml
    Storage buffer
    PBS/0,05% BSA/0,05% azide
  • BNUB0600-500
    BNUB0600-500
    500 μL
    500 µl
       
    Log in for availability
    1 * 500 µl
    1 * 500 µl - £491.00
    £491.00
    1 * 500 µl
       £491.00

    Supplier Article Number: BNUB0600-500
    Local Article Number: BTIUBNUB0600-500
     
     
    Specifications
    Antigen symbol
    CD2
    Antigen name
    CD2 molecule
    Environmentally Preferable
    Antigen synonyms
    Rosette receptor|Erythrocyte receptor|Ly-37|T-cell surface antigen T11/Leu-5|T11|T-cell surface antigen CD2|SRBC|LFA-3 receptor|LFA-2|Lymphocyte function antigen 2
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    Unconjugated
    Clone
    Clone LFA2/600
    Reactivity
    Primates, Pig, Human
    Host
    Mouse
    Gene ID
    523500
    Isotype
    IgG1 kappa
    ImmunoFluorescence
    Yes
    Concentration
    0,2 mg/ml
    Storage buffer
    PBS/0,05% BSA/0,05% azide