Northern Ireland
Order Entry
Anti-KRT14 Mouse Monoclonal Antibody (Biotin) [clone: KRT14/532]
Anti-KRT14 Mouse Monoclonal Antibody (Biotin) [clone: KRT14/532]
 :  Biotium
 
Anti-KRT14 Mouse Monoclonal Antibody (Biotin) [clone: KRT14/532]
 :  Biotium

 

  • Catalog No:
  • BNCB0532-100
  • BNCB0532-500
  • Antigen symbol:
  • KRT14
  • KRT14
  • Antigen name:
  • Cytokeratin 14
  • Cytokeratin 14
  • Antigen synonyms:
  • k14|ebs3|Dowling Meara|ebs4|NFJ|krt14|CK-14|Epidermolysis Bullosa Simplex|Keratin 14|Koebner|Keratin Type I Cytoskeletal 14
  • k14|ebs3|Dowling Meara|ebs4|NFJ|krt14|CK-14|Epidermolysis Bullosa Simplex|Keratin 14|Koebner|Keratin Type I Cytoskeletal 14
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Clone:
  • Clone KRT14/532
  • Clone KRT14/532
  • Reactivity:
  • Human, Rat, Mouse
  • Human, Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 6545380
  • 6545380
  • Isotype:
  • IgG3
  • IgG3
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

 

 

  •  
     
    Size
    Pk
     
     
  • BNCB0532-100
    BNCB0532-100
    100 μL
    100 µl
       
       
    1 * 100 µl
    1 * 100 µl - £195.00
    £195.00
    1 * 100 µl
       £195.00

     
     
    Specifications
    Antigen symbol
    KRT14
    Antigen name
    Cytokeratin 14
    Environmentally Preferable
    Antigen synonyms
    k14|ebs3|Dowling Meara|ebs4|NFJ|krt14|CK-14|Epidermolysis Bullosa Simplex|Keratin 14|Koebner|Keratin Type I Cytoskeletal 14
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    Biotin
    Clone
    Clone KRT14/532
    Reactivity
    Human, Rat, Mouse
    Host
    Mouse
    Gene ID
    6545380
    Isotype
    IgG3
    ImmunoChemistry
    Yes
    ImmunoFluorescence
    Yes
    Concentration
    0,1 mg/ml
    Storage buffer
    PBS/0,1% BSA/0,05% azide
  • BNCB0532-500
    BNCB0532-500
    500 μL
    500 µl
       
       
    1 * 500 µl
    1 * 500 µl - £538.00
    £538.00
    1 * 500 µl
       £538.00

     
     
    Specifications
    Antigen symbol
    KRT14
    Antigen name
    Cytokeratin 14
    Environmentally Preferable
    Antigen synonyms
    k14|ebs3|Dowling Meara|ebs4|NFJ|krt14|CK-14|Epidermolysis Bullosa Simplex|Keratin 14|Koebner|Keratin Type I Cytoskeletal 14
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    Biotin
    Clone
    Clone KRT14/532
    Reactivity
    Human, Rat, Mouse
    Host
    Mouse
    Gene ID
    6545380
    Isotype
    IgG3
    ImmunoChemistry
    Yes
    ImmunoFluorescence
    Concentration
    0,1 mg/ml
    Storage buffer
    PBS/0,1% BSA/0,05% azide