Anti-AFP Mouse Monoclonal Antibody (CF488A) [clone: C2]
: Biotium
This antibody recognizes an oncofetal glycoprotein with a single chain of 70 kDa, which is identified as alpha fetoprotein (AFP) (ISOBM TD-2 workshop). This MAb is highly specific to AFP and shows no cross-reaction with other oncofetal antigens or serum albumin. AFP is normally synthesized in the liver, intestinal tract, and yolk sac of the fetus. Antibody to AFP has been shown to be useful in detecting hepatocellular carcinomas (HCC) and germ cell neoplasms, especially yolk sac tumors.
Type: Primary
Antigen: AFP
Clonality: Monoclonal
Clone: C2
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse
- Catalog No:
- BNC880523-500
- BNC880523-100
- Antigen symbol:
- AFP
- AFP
- Antigen name:
- AFP
- AFP
- Antigen synonyms:
- FETA|Alpha fetoglobulin|HPAFP
- FETA|Alpha fetoglobulin|HPAFP
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 488A
- CF® 488A
- Clone:
- Clone C2
- Clone C2
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- 518808
- 518808
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide
- SizePk
Specifications
Antigen symbolAFPAntigen nameAFPEnvironmentally PreferableAntigen synonymsFETA|Alpha fetoglobulin|HPAFPAntibody typePrimaryClonalityMonoclonalConjugationCF® 488ACloneClone C2ReactivityHuman, MouseHostMouseGene ID518808IsotypeIgG1 kappaImmunoChemistryYesImmunoFluorescenceYesConcentration0,1 mg/mlStorage bufferPBS/0,1% BSA/0,05% azideSpecifications
Antigen symbolAFPAntigen nameAFPEnvironmentally PreferableAntigen synonymsFETA|Alpha fetoglobulin|HPAFPAntibody typePrimaryClonalityMonoclonalConjugationCF® 488ACloneClone C2ReactivityHuman, MouseHostMouseGene ID518808IsotypeIgG1 kappaImmunoChemistryYesImmunoFluorescenceYesConcentration0,1 mg/mlStorage bufferPBS/0,1% BSA/0,05% azide