About this item
Recognizes a cell surface glycoprotein of 80-95 kDa (CD44) on lymphocytes, monocytes, and granulocytes (Leucocyte Typing Workshop V). Its epitope is resistant to digestion by trypsin and chymotrypsin. This MAb selectively interferes with lymphocyte binding to lymph node, mucosal and synovial endothelium. The CD44 family of glycoproteins exists in a number of variant isoforms, the most common being the standard 85-95 kDa or hematopoietic variant (CD44s). Higher molecular weight isoforms are described in epithelial cells (CD44v), which are believed to function in intercellular adhesion and stromal binding. CD44 immunostaining is commonly used for the discrimination of urothelial transitional cell carcinoma in-situ from non-neoplastic changes in the urothelium.
Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: 156-3C11
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Baboon, Green monkey, Human
- Catalog No:
- BNC880460-500
- BNC880460-100
- Antigen symbol:
- CD44
- CD44
- Antigen name:
- CD44 Standard
- CD44 Standard
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 488A
- CF® 488A
- Clone:
- Clone 156-3C11
- Clone 156-3C11
- Reactivity:
- Baboon, Green monkey, Human
- Baboon, Green monkey, Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 502328
- 502328
- Isotype:
- IgG2a kappa
- IgG2a kappa
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide
Product Information
- Catalog #Supplier #SizePkAvailabilityPrice
- BNC880460-500BNC880460-500500 μL500 µl1 * 500 µl£538.00
Specifications
Antigen symbolCD44Antigen nameCD44 StandardEnvironmentally PreferableAntibody typePrimaryClonalityMonoclonalConjugationCF® 488ACloneClone 156-3C11ReactivityBaboon, Green monkey, HumanHostMouseGene ID502328IsotypeIgG2a kappaWestern blotYesImmunoChemistryYesImmunoFluorescenceConcentration0,1 mg/mlStorage bufferPBS/0,1% BSA/0,05% azide - BNC880460-100BNC880460-100100 μL100 µl1 * 100 µl£195.00
Specifications
Antigen symbolCD44Antigen nameCD44 StandardEnvironmentally PreferableAntibody typePrimaryClonalityMonoclonalConjugationCF® 488ACloneClone 156-3C11ReactivityBaboon, Green monkey, HumanHostMouseGene ID502328IsotypeIgG2a kappaWestern blotYesImmunoChemistryYesImmunoFluorescenceYesConcentration0,1 mg/mlStorage bufferPBS/0,1% BSA/0,05% azide