Northern Ireland
Order Entry
Anti-NEFH Mouse Monoclonal Antibody (CF405S) [clone: RT-97]
Anti-NEFH Mouse Monoclonal Antibody (CF405S) [clone: RT-97]
 :  Biotium
 
Anti-NEFH Mouse Monoclonal Antibody (CF405S) [clone: RT-97]
 :  Biotium

 

  • Catalog No:
  • BNC040454-500
  • BNC040454-100
  • Antigen symbol:
  • NEFH
  • NEFH
  • Antigen name:
  • Neurofilament, Heavy Polypeptide
  • Neurofilament, Heavy Polypeptide
  • Antigen synonyms:
  • Neurofilament H|Neurofilament Heavy Polypeptide 200kDa|NEFH|NF-H|Neurofilament Triplet H Protein|NF200
  • Neurofilament H|Neurofilament Heavy Polypeptide 200kDa|NEFH|NF-H|Neurofilament Triplet H Protein|NF200
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 405S
  • CF® 405S
  • Clone:
  • Clone RT-97
  • Clone RT-97
  • Reactivity:
  • Pig, Chicken, Human, Rat, Mouse
  • Pig, Chicken, Human, Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 198760
  • 198760
  • Isotype:
  • IgG1
  • IgG1
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

 

 

  •  
     
    Size
    Pk
     
     
  • BNC040454-500
    BNC040454-500
    500 μL
    500 µl
       
       
    1 * 500 µl
    1 * 500 µl - £538.00
    £538.00
    1 * 500 µl
       £538.00

     : BNC040454-500
     : BTIUBNC040454-500
     
     
    Specifications
    Antigen symbol
    NEFH
    Antigen name
    Neurofilament, Heavy Polypeptide
    Environmentally Preferable
    Antigen synonyms
    Neurofilament H|Neurofilament Heavy Polypeptide 200kDa|NEFH|NF-H|Neurofilament Triplet H Protein|NF200
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    CF® 405S
    Clone
    Clone RT-97
    Reactivity
    Pig, Chicken, Human, Rat, Mouse
    Host
    Mouse
    Gene ID
    198760
    Isotype
    IgG1
    ImmunoChemistry
    Yes
    ImmunoPrecipitation
    Yes
    ImmunoFluorescence
    Concentration
    0,1 mg/ml
    Storage buffer
    PBS/0,1% BSA/0,05% azide
  • BNC040454-100
    BNC040454-100
    100 μL
    100 µl
       
       
    1 * 100 µl
    1 * 100 µl - £195.00
    £195.00
    1 * 100 µl
       £195.00

     : BNC040454-100
     : BTIUBNC040454-100
     
     
    Specifications
    Antigen symbol
    NEFH
    Antigen name
    Neurofilament, Heavy Polypeptide
    Environmentally Preferable
    Antigen synonyms
    Neurofilament H|Neurofilament Heavy Polypeptide 200kDa|NEFH|NF-H|Neurofilament Triplet H Protein|NF200
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    CF® 405S
    Clone
    Clone RT-97
    Reactivity
    Pig, Chicken, Human, Rat, Mouse
    Host
    Mouse
    Gene ID
    198760
    Isotype
    IgG1
    ImmunoChemistry
    Yes
    ImmunoPrecipitation
    Yes
    ImmunoFluorescence
    Yes
    Concentration
    0,1 mg/ml
    Storage buffer
    PBS/0,1% BSA/0,05% azide