Anti-Biotin Mouse Monoclonal Antibody (CF405S) [clone: Hyb8]
: Biotium
This antibody recognizes both the free and protein-conjugated (either soluble or cell bound) form of biotin. This MAb is highly specific to biotin and shows no cross-reaction with other structurally related compounds. It has a very high affinity for biotin and is excellent for use in various amplification techniques. In some applications, localization of biotinylated probes with avidin produces unacceptably high background staining. Anti-biotin antibody may be substituted to decrease this noise.
Type: Primary
Antigen: Not Applicable
Clonality: Monoclonal
Clone: Hyb8
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: All species
- Catalog No:
- BNC040400-100
- BNC040400-500
- Antigen name:
- Biotin
- Biotin
- Antigen synonyms:
- Vitamin B7 or Vitamin H
- Vitamin B7 or Vitamin H
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 405S
- CF® 405S
- Clone:
- Clone Hyb8
- Clone Hyb8
- Reactivity:
- Multiple
- Multiple
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide
Product Information
- Catalog #Supplier #SizePkAvailabilityPrice
- BNC040400-100BNC040400-100100 μL100 µl1 * 100 µl£195.00
Specifications
Antigen nameBiotinEnvironmentally PreferableAntigen synonymsVitamin B7 or Vitamin HAntibody typePrimaryClonalityMonoclonalConjugationCF® 405SCloneClone Hyb8ReactivityMultipleHostMouseIsotypeIgG1Western blotYesImmunoChemistryYesImmunoFluorescenceYesConcentration0,1 mg/mlStorage bufferPBS/0,1% BSA/0,05% azide - BNC040400-500BNC040400-500500 μL500 µl1 * 500 µl£538.00
Specifications
Antigen nameBiotinEnvironmentally PreferableAntigen synonymsVitamin B7 or Vitamin HAntibody typePrimaryClonalityMonoclonalConjugationCF® 405SCloneClone Hyb8ReactivityMultipleHostMouseIsotypeIgG1Western blotYesImmunoChemistryYesImmunoFluorescenceYesConcentration0,1 mg/mlStorage bufferPBS/0,1% BSA/0,05% azide