About this item
PAb122 binds to the C-terminus (aa370-378) of both wild type and mutated p53. When microinjected into nuclei, PAb122 blocked re-entry into the S-phase of the cell cycle. Mutation and/or allelic loss of p53 is one of the causes of a variety of mesenchymal and epithelial tumors. If it occurs in the germ line, such tumors run in families. p53 Binds to a DNA consensus sequence, the p53 response element, and it regulates normal cell growth cycle events by activating transcription of genes, involved either in progression through the cycle, or causing arrest in G1 when the genome is damaged. In most transformed and tumor cells the concentration of p53 is increased 51000 fold over the minute concentrations (1000 molecules cell) in normal cells, principally due to the increased half-life (4 h) compared to that of the wild-type (20 min). p53 Localizes in the nucleus, but is detectable at the plasma membrane during mitosis and when certain mutations modulate cytoplasmic/nuclear distribution. p53 Is the most commonly mutated gene in spontaneously occurring human cancers. Mutations arise with an average frequency of 70% but incidence varies from zero in carcinoid lung tumors to 97% in primary melanomas. High concentrations of p53 protein are transiently expressed in human epidermis and superficial dermal fibroblasts following mild ultraviolet irradiation.
Type: Primary
Antigen: TP53
Clonality: Monoclonal
Clone: PAb122
Conjugation: CF640R
Epitope:
Host: Rabbit
Isotype: IgG2b kappa
Reactivity: Dog, Hamster, Human, Monkey, Mouse, Rat
- Catalog No:
- BNC400338-500
- BNC400338-100
- Antigen symbol:
- TP53
- TP53
- Antigen name:
- p53
- p53
- Antigen synonyms:
- Transformation Related Protein 53 (TRP53)|Tumor Protein p53|BCC7|Cellular Tumor Antigen p53|Tumor Suppressor p53|TP53|LFS1|Antigen NY-CO-13
- Transformation Related Protein 53 (TRP53)|Tumor Protein p53|BCC7|Cellular Tumor Antigen p53|Tumor Suppressor p53|TP53|LFS1|Antigen NY-CO-13
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 640R
- CF® 640R
- Clone:
- Clone PAb122
- Clone PAb122
- Reactivity:
- Hamster, Monkey, Human, Dog, Rat, Mouse
- Hamster, Monkey, Human, Dog, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 654481
- 654481
- Isotype:
- IgG2b kappa
- IgG2b kappa
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide
Product Information
- Catalog #Supplier #SizePkAvailabilityPrice
- BNC400338-500BNC400338-500500 μL500 µl1 * 500 µl£538.00
Specifications
Antigen symbolTP53Antigen namep53Environmentally PreferableAntigen synonymsTransformation Related Protein 53 (TRP53)|Tumor Protein p53|BCC7|Cellular Tumor Antigen p53|Tumor Suppressor p53|TP53|LFS1|Antigen NY-CO-13Antibody typePrimaryClonalityMonoclonalConjugationCF® 640RCloneClone PAb122ReactivityHamster, Monkey, Human, Dog, Rat, MouseHostRabbitGene ID654481IsotypeIgG2b kappaImmunoChemistryYesImmunoFluorescenceYesConcentration0,1 mg/mlStorage bufferPBS/0,1% BSA/0,05% azide - BNC400338-100BNC400338-100100 μL100 µl1 * 100 µl£195.00
Specifications
Antigen symbolTP53Antigen namep53Environmentally PreferableAntigen synonymsTransformation Related Protein 53 (TRP53)|Tumor Protein p53|BCC7|Cellular Tumor Antigen p53|Tumor Suppressor p53|TP53|LFS1|Antigen NY-CO-13Antibody typePrimaryClonalityMonoclonalConjugationCF® 640RCloneClone PAb122ReactivityHamster, Monkey, Human, Dog, Rat, MouseHostRabbitGene ID654481IsotypeIgG2b kappaImmunoChemistryYesImmunoFluorescenceYesConcentration0,1 mg/mlStorage bufferPBS/0,1% BSA/0,05% azide