Anti-ITGB2 Mouse Monoclonal Antibody (CF405M) [clone: 68-5A5]

Supplier: Biotium
BNC050321-100EA 179 GBP
BNC050321-100 BNC050321-500
Anti-ITGB2 Mouse Monoclonal Antibody (CF405M) [clone: 68-5A5]
This antibody recognizes a transmembrane glycoprotein of 95 kDa, identified as CD18 or integrin -2 (Workshop III). It complexes non-covalently with either L, M, or X integrin (CD11a, b, or c) to form the heterodimers, LFA-1, Mac-1, and p150,95, respectively. LFA-1 is the receptor for three members of the Ig supergene family of proteins, ICAM-1 (CD54), ICAM-2 (CD102), and Mac-1 and p150,95 bind to ICAM-1, fibrinogen, and iC3b. ICAM-3 (CD50). CD18/CD11 heterodimeric molecules are involved with cell/cell and cell/extracellular adhesion in immune and inflammatory responses. This MAb blocks these cellular interactions.

Type: Primary
Antigen: ITGB2
Clonality: Monoclonal
Clone: 68-5A5
Conjugation: CF405M
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR