Northern Ireland
Order Entry
Anti-Lewis A Mouse Monoclonal Antibody (Purified) [clone: 7LE]
 :  Biotium
 
Anti-Lewis A Mouse Monoclonal Antibody (Purified) [clone: 7LE]
 :  Biotium

 

  • Catalog No:
  • BNUB0311-500
  • BNUB0311-100
  • Antigen name:
  • Lewis A
  • Lewis A
  • Antigen synonyms:
  • Lewis A antigen
  • Lewis A antigen
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • Clone 7LE
  • Clone 7LE
  • Reactivity:
  • Human, Mouse
  • Human, Mouse
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,2 mg/ml
  • 0,2 mg/ml
  • Storage buffer:
  • PBS/0,05% BSA/0,05% azide
  • PBS/0,05% BSA/0,05% azide

 

Product Information

  • Catalog #
    Supplier #
    Size
    Pk
    Availability
    Price
  • BNUB0311-500
    BNUB0311-500
    500 μL
    500 µl
       
    Log in for availability
    1 * 500 µl
    1 * 500 µl - £491.00
    £491.00
    1 * 500 µl
       £491.00

     : BNUB0311-500
     : BTIUBNUB0311-500
     
     
    Specifications
    Antigen name
    Lewis A
    Environmentally Preferable
    Antigen synonyms
    Lewis A antigen
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    Unconjugated
    Clone
    Clone 7LE
    Reactivity
    Human, Mouse
    Host
    Mouse
    Isotype
    IgG1 kappa
    ImmunoChemistry
    Yes
    ImmunoPrecipitation
    Yes
    ImmunoFluorescence
    Yes
    Concentration
    0,2 mg/ml
    Storage buffer
    PBS/0,05% BSA/0,05% azide
  • BNUB0311-100
    BNUB0311-100
    100 μL
    100 µl
       
       
    1 * 100 µl
    1 * 100 µl - £228.00
    £228.00
    1 * 100 µl
       £228.00

     : BNUB0311-100
     : BTIUBNUB0311-100
     
     
    Specifications
    Antigen name
    Lewis A
    Environmentally Preferable
    Antigen synonyms
    Lewis A antigen
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    Unconjugated
    Clone
    Clone 7LE
    Reactivity
    Human, Mouse
    Host
    Mouse
    Isotype
    IgG1 kappa
    ImmunoChemistry
    Yes
    ImmunoPrecipitation
    Yes
    ImmunoFluorescence
    Yes
    Concentration
    0,2 mg/ml
    Storage buffer
    PBS/0,05% BSA/0,05% azide