Northern Ireland
Order Entry
Anti-CD7 Mouse Monoclonal Antibody (CF405S) [clone: B-F12]
 :  Biotium
 
Anti-CD7 Mouse Monoclonal Antibody (CF405S) [clone: B-F12]
 :  Biotium

 

  • Catalog No:
  • BNC040310-100
  • BNC040310-500
  • Antigen symbol:
  • CD7
  • CD7
  • Antigen synonyms:
  • TP41|Leu9|T-cell surface antigen Leu-9|T-cell leukemia antigen|Tp40|p41|GP40
  • TP41|Leu9|T-cell surface antigen Leu-9|T-cell leukemia antigen|Tp40|p41|GP40
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 405S
  • CF® 405S
  • Clone:
  • Clone B-F12
  • Clone B-F12
  • Reactivity:
  • Pig, Human, Guinea pig
  • Pig, Human, Guinea pig
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 186820
  • 186820
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

 

Product Information

  • Catalog #
    Supplier #
    Size
    Pk
    Availability
    Price
  • BNC040310-100
    BNC040310-100
    100 μL
    100 µl
       
    Log in for availability
    1 * 100 µl
    1 * 100 µl - £195.00
    £195.00
    1 * 100 µl
       £195.00

     
     
    Specifications
    Antigen symbol
    CD7
    Environmentally Preferable
    Antigen synonyms
    TP41|Leu9|T-cell surface antigen Leu-9|T-cell leukemia antigen|Tp40|p41|GP40
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    CF® 405S
    Clone
    Clone B-F12
    Reactivity
    Pig, Human, Guinea pig
    Host
    Mouse
    Gene ID
    186820
    Isotype
    IgG2a kappa
    ImmunoFluorescence
    Yes
    Concentration
    0,1 mg/ml
    Storage buffer
    PBS/0,1% BSA/0,05% azide
  • BNC040310-500
    BNC040310-500
    500 μL
    500 µl
       
    Log in for availability
    1 * 500 µl
    1 * 500 µl - £538.00
    £538.00
    1 * 500 µl
       £538.00

     
     
    Specifications
    Antigen symbol
    CD7
    Environmentally Preferable
    Antigen synonyms
    TP41|Leu9|T-cell surface antigen Leu-9|T-cell leukemia antigen|Tp40|p41|GP40
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    CF® 405S
    Clone
    Clone B-F12
    Reactivity
    Pig, Human, Guinea pig
    Host
    Mouse
    Gene ID
    186820
    Isotype
    IgG2a kappa
    ImmunoFluorescence
    Yes
    Concentration
    0,1 mg/ml
    Storage buffer
    PBS/0,1% BSA/0,05% azide