About this item
Laminins are large hetero-trimeric, non-collagenous glycoproteins composed of α, β, and γ chains. This MAb reacts with laminin B2/1 chain of ~210 kDa and does not cross-react with other basement membrane components or fibronectin. Its specificity was established by immunoprecipitation and immunofluorescence of human skeletal muscle and kidney with laminin chain-specific MAbs. Epithelial sheets in vivo are separated from the mesenchymal elements of the stroma by a thin layer of a specialized type of extracellular matrix termed the basement membrane (BM). This structure consists of individual components, some of which are ubiquitous in BMs and some are not. The ubiquitous ones comprise laminin (LN), entactin/nidogen (EN), collagen type IV (CIV), and large heparan sulfate proteoglycan (HSPG), which interact specifically with each other to form a continuous and regular BM. Alterations of BM integrity, from local discontinuities up to complete loss, are described in many types of human and animal epithelial neoplasms. This MAb stains uniformly all human and murine basement membranes.
Type: Primary
Antigen: LAMC1
Clonality: Monoclonal
Clone: A5
Conjugation: CF568
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Human, Mouse
Specifications
- Catalog No:
- BNC680308-500
- BNC680308-100
- Antigen symbol:
- LAMC1
- LAMC1
- Antigen name:
- Laminin
- Laminin
- Antigen synonyms:
- Laminin B2 chain|LAMC1|Laminin gamma 1 chain|LAMB2|Laminin gamma 1
- Laminin B2 chain|LAMC1|Laminin gamma 1 chain|LAMB2|Laminin gamma 1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 568
- CF® 568
- Clone:
- Clone A5
- Clone A5
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rat
- Rat
- Gene ID:
- 609663
- 609663
- Isotype:
- IgG2a kappa
- IgG2a kappa
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide
Specifications
Product Information
- Catalog #Supplier #SizePkAvailabilityPrice
- BNC680308-500BNC680308-500500 μL500 µl1 * 500 µl£538.00: BNC680308-500: BTIUBNC680308-500
Specifications
Antigen symbolLAMC1Antigen nameLamininEnvironmentally PreferableAntigen synonymsLaminin B2 chain|LAMC1|Laminin gamma 1 chain|LAMB2|Laminin gamma 1Antibody typePrimaryClonalityMonoclonalConjugationCF® 568CloneClone A5ReactivityHuman, MouseHostRatGene ID609663IsotypeIgG2a kappaImmunoChemistryYesImmunoFluorescenceYesConcentration0,1 mg/mlStorage bufferPBS/0,1% BSA/0,05% azide - : BNC680308-100: BTIUBNC680308-100
Specifications
Antigen symbolLAMC1Antigen nameLamininEnvironmentally PreferableAntigen synonymsLaminin B2 chain|LAMC1|Laminin gamma 1 chain|LAMB2|Laminin gamma 1Antibody typePrimaryClonalityMonoclonalConjugationCF® 568CloneClone A5ReactivityHuman, MouseHostRatGene ID609663IsotypeIgG2a kappaImmunoChemistryYesImmunoFluorescenceYesConcentration0,1 mg/mlStorage bufferPBS/0,1% BSA/0,05% azide