Anti-Phosphotyrosine Mouse Monoclonal Antibody (HRP) [clone: PY20]

Supplier: Biotium
BNCH0232-500EA 468 GBP
BNCH0232-500 BNCH0232-100
Anti-Phosphotyrosine Mouse Monoclonal Antibody (HRP) [clone: PY20]
Protein phosphorylation is a fundamental event in the regulation of a large number of intracellular processes. Phosphorylation of specific tyrosine residues is the result of activation or stimulation of their respective protein tyrosine kinases. The phosphorylated proteins can be auto-phosphorylated kinases or certain cellular protein substrates. Tyrosine-phosphorylated proteins are involved in signal transduction and in the regulation of cell proliferation. Antibody to phosphotyrosine provides an excellent tool for the detection, characterization, and purification of phosphotyrosine containing proteins. This MAb shows no cross-reaction with other phosphoamino acids and is superb for multiple applications including staining of formalin/paraffin tissues.

Type: Primary
Antigen: Not Applicable
Clonality: Monoclonal
Clone: PY20
Conjugation: HRP
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: All species
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR