Anti-CMV-p65 Mouse Monoclonal Antibody (CF405S) [clone: CMV101]
: Biotium
Ten to forty percent of the patients with acquired immunodeficiency syndrome (AIDS) develop cytomegalovirus (CMV) infections. In some patients with AIDS, CMV is detected in the bronchoalveolar lavage fluid (BALF), urine, and other specimens, even when there are no symptoms of CMV disease. An indicator of active CMV infection is needed to facilitate the diagnosis of CMV disease in patients with AIDS or HIV infection. CMV p65 antigen was detected in the leukocytes of both the peripheral blood and BALF during the early phase of CMV disease.
Type: Primary
Antigen: Not Applicable
Clonality: Monoclonal
Clone: CMV101
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: CMV
- Catalog No:
- BNC040101-500
- BNC040101-100
- Antigen name:
- CMV-p65
- CMV-p65
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 405S
- CF® 405S
- Clone:
- Clone CMV101
- Clone CMV101
- Reactivity:
- Cytomegalovirus
- Cytomegalovirus
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide
- SizePk
- : BNC040101-500: BTIUBNC040101-500
Specifications
Antigen nameCMV-p65Environmentally PreferableAntibody typePrimaryClonalityMonoclonalConjugationCF® 405SCloneClone CMV101ReactivityCytomegalovirusHostMouseIsotypeIgG1 kappaELISAYesImmunoChemistryYesImmunoFluorescenceYesConcentration0,1 mg/mlStorage bufferPBS/0,1% BSA/0,05% azide - : BNC040101-100: BTIUBNC040101-100
Specifications
Antigen nameCMV-p65Environmentally PreferableAntibody typePrimaryClonalityMonoclonalConjugationCF® 405SCloneClone CMV101ReactivityCytomegalovirusHostMouseIsotypeIgG1 kappaELISAYesImmunoChemistryYesImmunoFluorescenceYesConcentration0,1 mg/mlStorage bufferPBS/0,1% BSA/0,05% azide