Northern Ireland
Order Entry
Anti-PGF Mouse Monoclonal Antibody (CF488A) [clone: PLGF93]
Anti-PGF Mouse Monoclonal Antibody (CF488A) [clone: PLGF93]
 :  Biotium
 
Anti-PGF Mouse Monoclonal Antibody (CF488A) [clone: PLGF93]
Supplier:  Biotium

 

  • Catalog No:
  • BNC880093-500
  • BNC880093-100
  • Antigen symbol:
  • PGF
  • PGF
  • Antigen name:
  • Placental growth factor
  • Placental growth factor
  • Antigen synonyms:
  • PIGF|PGFL|placental growth factor (PGF)|Vascular endothelial growth factor related protein
  • PIGF|PGFL|placental growth factor (PGF)|Vascular endothelial growth factor related protein
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 488A
  • CF® 488A
  • Clone:
  • Clone PLGF93
  • Clone PLGF93
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 252820
  • 252820
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ELISA:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

 

Product Information

  • Catalog #
    Supplier #
    Size
    Pk
    Availability
    Price
  • BNC880093-500
    BNC880093-500
    500 μL
    500 µl
       
    Log in for availability
    1 * 500 µl
    1 * 500 µl - £538.00
    £538.00
    1 * 500 µl
       £538.00

     
     
    Specifications
    Antigen symbol
    PGF
    Antigen name
    Placental growth factor
    Environmentally Preferable
    Antigen synonyms
    PIGF|PGFL|placental growth factor (PGF)|Vascular endothelial growth factor related protein
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    CF® 488A
    Clone
    Clone PLGF93
    Reactivity
    Human
    Host
    Mouse
    Gene ID
    252820
    Isotype
    IgG1 kappa
    ELISA
    Yes
    ImmunoFluorescence
    Yes
    Concentration
    0,1 mg/ml
    Storage buffer
    PBS/0,1% BSA/0,05% azide
  • BNC880093-100
    BNC880093-100
    100 μL
    100 µl
       
    Log in for availability
    1 * 100 µl
    1 * 100 µl - £195.00
    £195.00
    1 * 100 µl
       £195.00

     
     
    Specifications
    Antigen symbol
    PGF
    Antigen name
    Placental growth factor
    Environmentally Preferable
    Antigen synonyms
    PIGF|PGFL|placental growth factor (PGF)|Vascular endothelial growth factor related protein
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    CF® 488A
    Clone
    Clone PLGF93
    Reactivity
    Human
    Host
    Mouse
    Gene ID
    252820
    Isotype
    IgG1 kappa
    ELISA
    Yes
    ImmunoFluorescence
    Yes
    Concentration
    0,1 mg/ml
    Storage buffer
    PBS/0,1% BSA/0,05% azide