About this item
Recognizes a polypeptide which is identified as insulin, a 51-amino acid polypeptide composed of A and B chains connected through the C-peptide. Proinsulin, which has very little biological activity, is cleaved by proteases within its cell of origin into the insulin molecule and the C-terminal basic residue. Insulin enhances membrane transport of glucose, amino acids, and certain ions. It also promotes glycogen storage, formation of triglycerides, and synthesis of proteins and nucleic acids. Deficiency of insulin results in diabetes mellitus. The main storage site for insulin is the pancreatic islets. Antibodies to insulin are important as beta-cell and insulinoma marker.
Type: Primary
Antigen: INS
Clonality: Monoclonal
Clone: 2D11-H5
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Cow, Human, Mouse, Pig
Specifications
- Catalog No:
- BNC040030-500
- BNC040030-100
- Antigen symbol:
- INS
- INS
- Antigen name:
- Insulin
- Insulin
- Antigen synonyms:
- MODY10|ILPR|IDDM2|Proinsulin|IRDN
- MODY10|ILPR|IDDM2|Proinsulin|IRDN
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 405S
- CF® 405S
- Clone:
- Clone 2D11-H5
- Clone 2D11-H5
- Reactivity:
- Pig, Human, Bovine, Mouse
- Pig, Human, Bovine, Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- 272259
- 272259
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide
Specifications
Product Information
- Catalog #Supplier #SizePkAvailabilityPrice
- BNC040030-500BNC040030-500500 μL500 µl1 * 500 µl£538.00: BNC040030-500: BTIUBNC040030-500
Specifications
Antigen symbolINSAntigen nameInsulinEnvironmentally PreferableAntigen synonymsMODY10|ILPR|IDDM2|Proinsulin|IRDNAntibody typePrimaryClonalityMonoclonalConjugationCF® 405SCloneClone 2D11-H5ReactivityPig, Human, Bovine, MouseHostMouseGene ID272259IsotypeIgG1 kappaImmunoChemistryYesImmunoFluorescenceYesConcentration0,1 mg/mlStorage bufferPBS/0,1% BSA/0,05% azide - : BNC040030-100: BTIUBNC040030-100
Specifications
Antigen symbolINSAntigen nameInsulinEnvironmentally PreferableAntigen synonymsMODY10|ILPR|IDDM2|Proinsulin|IRDNAntibody typePrimaryClonalityMonoclonalConjugationCF® 405SCloneClone 2D11-H5ReactivityPig, Human, Bovine, MouseHostMouseGene ID272259IsotypeIgG1 kappaImmunoChemistryYesImmunoFluorescenceYesConcentration0,1 mg/mlStorage bufferPBS/0,1% BSA/0,05% azide