This MAb reacts with a wide variety of simple epithelia. It does not react with stratified squamous epithelia. It reacts with epithelial tumors of the gastrointestinal tract, lung, breast, pancreas, ovary, and thyroid. Cytokeratin 18, which belongs to the type A (acidic) subfamily of low molecular weight keratins, exists in combination with cytokeratin 8. It is reported that tissues from gastrointestinal tract are positive for both cytokeratin 8 and 18 but do not contain cytokeratin 14. Tissues from gastrointestinal tract, respiratory tract and urogenital tract, as well as endocrine and exocrine tissues and mesothelial cells are positive for cytokeratin 18.
Type: Primary
Antigen: KRT18
Clonality: Monoclonal
Clone: DC10
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
- Catalog No:
- BNUB0021-100
- BNUB0021-500
- Antigen symbol:
- KRT18
- KRT18
- Antigen name:
- Cytokeratin 18
- Cytokeratin 18
- Antigen synonyms:
- K18|KRT18|CYK18 Cytokeratin Endo B|CK18|Keratin-18|Kerd|Cell Proliferation-inducing Gene 46 Protein
- K18|KRT18|CYK18 Cytokeratin Endo B|CK18|Keratin-18|Kerd|Cell Proliferation-inducing Gene 46 Protein
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- Clone DC10
- Clone DC10
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 406013
- 406013
- Isotype:
- IgG1
- IgG1
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,2 mg/ml
- 0,2 mg/ml
- Storage buffer:
- PBS/0,05% BSA/0,05% azide
- PBS/0,05% BSA/0,05% azide
- SizePk
Specifications
Antigen symbolKRT18Antigen nameCytokeratin 18Environmentally PreferableAntigen synonymsK18|KRT18|CYK18 Cytokeratin Endo B|CK18|Keratin-18|Kerd|Cell Proliferation-inducing Gene 46 ProteinAntibody typePrimaryClonalityMonoclonalConjugationUnconjugatedCloneClone DC10ReactivityHumanHostMouseGene ID406013IsotypeIgG1Western blotYesImmunoChemistryYesImmunoFluorescenceYesConcentration0,2 mg/mlStorage bufferPBS/0,05% BSA/0,05% azideSpecifications
Antigen symbolKRT18Antigen nameCytokeratin 18Environmentally PreferableAntigen synonymsK18|KRT18|CYK18 Cytokeratin Endo B|CK18|Keratin-18|Kerd|Cell Proliferation-inducing Gene 46 ProteinAntibody typePrimaryClonalityMonoclonalConjugationUnconjugatedCloneClone DC10ReactivityHumanHostMouseGene ID406013IsotypeIgG1Western blotYesImmunoChemistryYesImmunoFluorescenceYesConcentration0,2 mg/mlStorage bufferPBS/0,05% BSA/0,05% azide